Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32809

Supelco

Desogestrel

VETRANAL®, analytical standard

Synonym(s):

13-Ethyl-11-methylene-18,19-dinor-17α-4-pregnen-20-yn-17-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

InChI

1S/C22H30O/c1-4-21-14-15(3)20-17-9-7-6-8-16(17)10-11-18(20)19(21)12-13-22(21,23)5-2/h2,8,17-20,23H,3-4,6-7,9-14H2,1H3/t17-,18-,19-,20+,21-,22-/m0/s1

InChI key

RPLCPCMSCLEKRS-BPIQYHPVSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

Desogestrel is a novel progestational steroid developed for utilization in oral contraceptives.

Application

Desogestrel may be used as an analytical reference standard for the determination of the analyte in:
  • Contraceptives and river water samples by high performance liquid chromatography/ion-trap mass spectrometry (LC-IT MS).
  • Pharmaceutical samples by LC-MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the sensitivity of miniaturized liquid chromatography-electrospray ionization-mass spectrometry for pharmaceutical analysis.
Kranendijk M, et al.
Journal of Separation Science, 28(14), 1796-1802 (2005)
Gabriele S Merki-Feld et al.
Cephalalgia : an international journal of headache, 33(5), 340-346 (2013-01-09)
Migraine seems to be substantially related to hormonal changes. However, migraine, but also the use of combined oral contraceptives (COC), is associated with an increased risk for vascular events. Therefore progestagen-only contraception is an important alternative to combined preparations. Clinical
High performance liquid chromatography/ion-trap mass spectrometry for separation and simultaneous determination of ethynylestradiol, gestodene, levonorgestrel, cyproterone acetate and desogestrel.
Matejicek D and Kuban V
Analytica Chimica Acta, 588(2), 304-315 (2007)
Tessa Madden et al.
Obstetrics and gynecology, 120(5), 1053-1059 (2012-10-24)
To estimate whether women who receive an immediate postabortion etonogestrel subdermal implant have higher discontinuation rates than women who undergo an interval insertion (insertion not associated with a recent pregnancy). This was a secondary analysis of the Contraceptive CHOICE Project.
Petra M Casey et al.
Contraception, 87(3), 370-374 (2012-09-11)
Bleeding irregularities represent the most common etonogestrel subdermal implant (ESI) removal indication. ESI placements (n=304) from June 2007 to April 2011 were grouped by removal indications. Group characteristics were compared using one-way analysis of variance, Kruskal-Wallis and χ(2) test. Of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service