Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32345

Supelco

Gatifloxacin sesquihydrate

VETRANAL®, analytical standard

Synonym(s):

(±)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O4 · 1.5H2O
CAS Number:
Molecular Weight:
402.42
Beilstein:
14390028
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O=C(O)C1=CN(C2CC2)C3=C(OC)C(N4CCNC(C)C4)=C(F)C=C3C1=O.O=C(O)C5=CN(C6CC6)C7=C(OC)C(N8CCNC(C)C8)=C(F)C=C7C5=O.O.O.O

InChI

1S/2C19H22FN3O4.3H2O/c2*1-10-8-22(6-5-21-10)16-14(20)7-12-15(18(16)27-2)23(11-3-4-11)9-13(17(12)24)19(25)26;;;/h2*7,9-11,21H,3-6,8H2,1-2H3,(H,25,26);3*1H2

InChI key

RMJMZKDEVNTXHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Luisa C Domingos et al.
Journal of pharmacological and toxicological methods, 83, 87-93 (2016-10-30)
The fourth-generation fluoroquinolones are widely used as ophthalmic antimicrobials. This study aimed to validate a new analytical technique for simultaneous quantification of gatifloxacin, moxifloxacin, and besifloxacin concentrations in the cornea and aqueous humor by liquid chromatography (LC) coupled to quadrupole
Patricia Marcianes et al.
AAPS PharmSciTech, 21(1), 15-15 (2019-12-07)
Tuberculosis (TB) is an infectious disease caused by Mycobacterium tuberculosis and represents one of the leading causes of mortality worldwide due to multidrug-resistant TB (MDR-TB). In our work, a new formulation of biodegradable PLGA microparticles was developed for pulmonary administration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service