Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31298

Supelco

Dimethyl terephthalate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1107185
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

1.04 (vs air)

vapor pressure

1.15 mmHg ( 93 °C)

Assay

≥98% (HPLC)

form

powder, crystals or chunks ((pellets))

autoignition temp.

1058 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

139-141 °C (lit.)

application(s)

environmental

format

neat

SMILES string

COC(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3

InChI key

WOZVHXUHUFLZGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

309.2 °F - (External MSDS)

Flash Point(C)

154 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnaud Lavalette et al.
Biomacromolecules, 3(2), 225-228 (2002-03-13)
The polymerization of dimethyl terephthalate and diethylene glycol by enzymic catalysis in toluene is described. The potential pi-pi interactions of the aromatic rings together with the relative flexibility of the diol segment have been regarded as synergy factors responsible for
[Hygienic evaluation of the allergic effect of dimethyl terephthalate after inhalation and oral administration].
G I Vinogradov et al.
Gigiena i sanitariia, (5)(5), 7-10 (1986-05-01)
R I Goncharova et al.
TSitologiia i genetika, 36(1), 14-25 (2002-05-16)
In the mouse transplacental test, EMS induced micronuclei and disturbed haemopoiesis in female bone marrow and foetal liver. Dimethyl terephthalate at the tested dose was ineffective in pregnant females increasing however the level of these events in foetuses. Hence, both
R I Goncharova et al.
Genetika, 24(7), 1226-1233 (1988-07-01)
Mutagenic activity of dimethyl terephthalate (DMtP) was evaluated using the bone marrow micronucleus test in mice. Clear clastogenic effect with the highest response in 24 h after a single i.p. injection was obtained at all concentrations used (0.2-1.0 mM/kg). The
Claudio A Téllez et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(8-9), 2171-2180 (2004-07-14)
Fourier transform infrared and Raman spectra of dimethylterephthalate (DMT), as microcrystalline powder, have been investigated. The vibrational spectra were calculated using the AM1 and PM3 semi empirical procedures, and the Møller-Plesset (MP2/DZV), and the Becke-Lee, Yang and Parr gradient-corrected correlation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service