Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14131

Supelco

Iopanoic acid

analytical standard

Synonym(s):

2-(3-Amino-2,4,6-triiodobenzyl)butyric acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropanoic acid, Iodopanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12I3NO2
CAS Number:
Molecular Weight:
570.93
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-156 °C

application(s)

clinical

format

neat

storage temp.

2-8°C

SMILES string

O=C(O)C(CC)CC1=C(I)C(N)=C(I)C=C1I

InChI

1S/C11H12I3NO2/c1-2-5(11(16)17)3-6-7(12)4-8(13)10(15)9(6)14/h4-5H,2-3,15H2,1H3,(H,16,17)

InChI key

OIRFJRBSRORBCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iopanoic acid is an oral cholecystographic agent, which is rich in iodine and is a potent inhibitor of 5′-deiodinase.
This radiographic contrast agent prevents the release of thyroid hormone from the thyroid gland. It also inhibits the conversion of thyroxine to triiodothyronine.

Application

Iopanoic acid may be used as a standard to investigate the presence and regulation of type 3 iodothyronine deiodinase in human cardiomyocytes.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Iopanoic acid rapidly controls type I amiodarone-induced thyrotoxicosis prior to thyroidectomy.
Bogazzi F, et al.
Journal of Endocrinological Investigation, 25(2), 176-180 (2002)
Evidence against a major role of L-thyroxine at the pituitary level: studies in rats treated with iopanoic acid (telepaque).
Obregon JM, et al.
Endocrinology, 106(6), 1827-1836 (1980)
Type 3 iodothyronine deiodinase is expressed in human induced pluripotent stem cell derived cardiomyocytes.
Nishimura K, et al.
Life Sciences (2018)
Xuehua Piao et al.
The Journal of allergy and clinical immunology, 143(1), 213-228 (2018-03-30)
A delicate balance between cell death and keratinocyte proliferation is crucial for normal skin development. Previous studies have reported that cellular FLICE (FADD-like ICE)-inhibitory protein plays a crucial role in prevention of keratinocytes from TNF-α-dependent apoptosis and blocking of dermatitis.
Janet L Markman et al.
Nature communications, 11(1), 1613-1613 (2020-04-03)
In men, the incidence of melanoma rises rapidly after age 50, and nearly two thirds of melanoma deaths are male. The immune system is known to play a key role in controlling the growth and spread of malignancies, but whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service