Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07589

Supelco

α-Mangostin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O6
CAS Number:
Molecular Weight:
410.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

COc1c(O)cc2Oc3cc(O)c(C\C=C(\C)C)c(O)c3C(=O)c2c1C\C=C(/C)C

InChI

1S/C24H26O6/c1-12(2)6-8-14-16(25)10-19-21(22(14)27)23(28)20-15(9-7-13(3)4)24(29-5)17(26)11-18(20)30-19/h6-7,10-11,25-27H,8-9H2,1-5H3

InChI key

GNRIZKKCNOBBMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Xanthone derivative isolated from mangosteen (Garcinia mangostana). Antioxidant and anti-inflammatory.
Xanthone derivative isolated from mangosteen (Garcinia mangostana). Antioxidant and anti-inflammatory. α-mangostin has been shown to induce apoptosis via the mitochondrial pathway, reduce cell proliferation and inhibit tumorigenesis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service