Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02150595

Osthole

primary reference standard

Synonym(s):

7-Methoxy-8-(3-methyl-2-butenyl)coumarin, 7-Methoxy-8-isopentenylcoumarin, Osthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O3
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

COc1ccc2C=CC(=O)Oc2c1C\C=C(\C)C

InChI

1S/C15H16O3/c1-10(2)4-7-12-13(17-3)8-5-11-6-9-14(16)18-15(11)12/h4-6,8-9H,7H2,1-3H3

InChI key

MBRLOUHOWLUMFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Active constituent of Cnidium monnieri, a traditional Chinese medicine used for its anti-inflammatory, analgesic, and anti-lipedemic activities. Dramatically decreased lipid accumulation in a quail model. Its neuroprotective effect on MPP(+)-induced cytotoxicity in PC12 cells supports the use of osthole as a therapeutic agent for the treatment of neurodegenerative disorders. Suppression of fatty acid synthase expression in HER2-overexpressing breast cancer cells indicates osthole as a promising agent for chemotherapy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ola D Alabi et al.
Life sciences, 102(2), 105-110 (2014-03-25)
Osthole, a coumarin derivative, has been used in Chinese medicine and studies have suggested a potential use in treatment of diabetes and cancers. Therefore, we investigated the effects of osthole and other coumarins on GLUT1 activity in two cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service