Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00130595

trans-Anethole

primary reference standard

Synonym(s):

4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
774229
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard
primary reference standard

form

crystals (white)
liquid (above 23 oC)

manufacturer/tradename

HWI

refractive index

n20/D 1.561 (lit.)

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Dolores López et al.
Chemico-biological interactions, 229, 36-43 (2015-02-01)
The profitable insecticidal action of monoterpenoids prompted us to test their efficiency against stored-grain beetle species, via inhibition of acetylcholinesterase (AChE). For this, we first studied the ability of the monoterpenoids geraniol, linalool, camphor, fenchone, carvone and γ-terpinene, besides the
Tomohiro Nabekura et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 70, 140-145 (2015-03-18)
Drug efflux transporter P-glycoprotein plays an important role in cancer chemotherapy. The nuclear factor-κB (NF-κB) transcription factors play critical roles in development and progression of cancer. In this study, the effects of natural compounds that can inhibit NF-κB activation on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service