Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-0116

Sigma-Aldrich

pH standard solution

pH 6.86 (25 °C), JCSS

Synonym(s):

pH 6.86 Buffer Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

Agency

JCSS

Quality Level

availability

available only in Japan

pH

6.86 (25 °C)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service