Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37-0530

Sigma-Aldrich

Zinc Standard for ICP

suitable for atomic absorption spectrometry, 1000 ppm Zn+2, 1 mg/mL Zn+2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

form

liquid

availability

available only in Japan

concentration

1 mg/mL Zn+2
1000 ppm Zn+2

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service