Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1440

Sigma-Aldrich

Silver sulfate

JIS special grade, ≥99.5%

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.5%

form

solid

availability

available only in Japan

mp

652 °C (lit.)

storage temp.

15-25°C

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service