Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-1030

Sigma-Aldrich

Ammonium iron(III) citrate

CP, green

Synonym(s):

Ammonium ferric citrate, Ferric ammonium citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H8O7 ·xFe3+ · yNH3
CAS Number:
UNSPSC Code:
12352300

grade

CP

form

solid

availability

available only in Japan

concentration

13.5-17.0% Fe

color

green

storage temp.

15-25°C

SMILES string

N.OC12CC(=O)O[Fe](OC(=O)C1)OC(=O)C2

InChI

1S/C6H8O7.Fe.H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H3/q;+3;/p-3

InChI key

FRHBOQMZUOWXQL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan M Mitchell et al.
Neurobiology of aging, 32(6), 1114-1123 (2009-06-30)
HFE gene variants are relatively common genetic variants in Caucasians. The H63D HFE genetic variant has been repeatedly associated with a number of neurodegenerative diseases. We developed neuroblastoma cell lines expressing different HFE polymorphisms to explore the mechanisms behind these
Yinghui Zhang et al.
Antioxidants & redox signaling, 13(7), 999-1009 (2010-04-22)
Cytosolic ferritins sequester and store iron, consequently protecting cells against iron-mediated free radical damage. However, the mechanisms of iron exit from the ferritin cage and reutilization are largely unknown. In a previous study, we found that mitochondrial ferritin (MtFt) expression
Tomoya Ohno et al.
Journal of biochemistry, 151(1), 109-114 (2011-10-14)
There is a little information about the effects of iron overload on cartilage metabolism. In the present study, we examined the effects of excess iron on the differentiation and mineralization of cultured chondrocytes, ATDC5 cells. We used ferric ammonium citrate
Rita Polati et al.
Journal of proteomics, 76 Spec No., 10-27 (2012-07-28)
Macrophages play a critical role at the crossroad between iron metabolism and immunity, being able to store and recycle iron derived from the phagocytosis of senescent erythrocytes. The way by which macrophages manage non-heme iron at physiological concentration is still
Yulia Pollak et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(1), 37-48 (2012-03-27)
Iron accumulation and iron-related oxidative stress are involved in several pathological conditions and provide a rationale for the development of iron chelators as novel promising therapeutic strategies. Thus, we have recently synthesized multifunctional non-toxic, brain permeable iron chelating compounds, M30

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service