Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-2020

Sigma-Aldrich

Celite® 545

CP

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhui Xiong et al.
Journal of colloid and interface science, 368(1), 528-532 (2011-12-14)
This paper presents a novel technique integrating bulk-sensitive and surface-sensitive XANES methods to distinguish between physisorption and chemisorption for phosphate adsorption onto ferrihydrite-modified diatomite (FHMD). XANES P K-edge, L-edge, and Fe M-edge spectra were obtained for reference samples (K(2)HPO(4) and
Xiao-Li Yang et al.
Bioresource technology, 101(23), 9178-9184 (2010-08-06)
This paper examined the effect of diatomite addition on membrane fouling and process performance in an anoxic/oxic submerged membrane bioreactor (A/O MBR). Particle size distribution, molecular weight distribution and microbial activity have been investigated to characterize the sludge mixed liquor.
Dayvson J Palmeira et al.
Molecules (Basel, Switzerland), 16(11), 9697-9713 (2011-11-25)
Lipase-catalyzed kinetic resolution of aryltrimethylsilyl chiral alcohols through a transesterification reaction was studied. The optimal conditions found for the kinetic resolution of m- and p-aryltrimethylsilyl chiral alcohols, led to excellent results, high conversions (c = 50%), high enantiomeric ratios (E
Gang Pan et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(8), 2205-2212 (2012-01-25)
The formation mechanism of a nanoscale gas state is studied on inorganic clay surfaces modified with hexamethyldisilazane, which show different contact angles in ethanol-water solutions. As the dissolved oxygen becomes oversaturated due to the decrease in ethanol-water ratio, oxygen nanoscale
Farhana Maqbool et al.
Journal of hazardous materials, 237-238, 262-269 (2012-09-15)
The present study reports the effect of bioaugmentation by free and immobilized bacterial culture on the rhizodegradation of petroleum-polluted soil using Sesbania cannabina plant. Total petroleum hydrocarbon (TPH), hydrocarbon-degrading bacterial counts, microbial activity and root morphology were assessed during 120

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service