Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1060

Sigma-Aldrich

Calcium stearate

CP, 6.2-7.2% Ca basis

Synonym(s):

Octadecanoic acid calcium salt, Stearic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COO]2Ca
CAS Number:
Molecular Weight:
607.02
Beilstein:
3919698
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

CP

form

solid

composition

Ca, 6.2-7.2%

availability

available only in Japan

storage temp.

15-25°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Ca]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Ca/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

CJZGTCYPCWQAJB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service