Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-6070

Sigma-Aldrich

Antimony(V) chloride

Synonym(s):

Antimony pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl5
CAS Number:
Molecular Weight:
299.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor density

>10.2 (vs air)

vapor pressure

0.13 psi ( 55 °C)

form

liquid

reaction suitability

reagent type: catalyst
core: antimony

availability

available only in Japan

bp

92 °C/30 mmHg (lit.)

mp

2.8 °C (lit.)

density

2.36 g/mL at 25 °C (lit.)

SMILES string

Cl[Sb](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Sb/h5*1H;/q;;;;;+5/p-5

InChI key

VMPVEPPRYRXYNP-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Suitability

PCB analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-transit chemical spill--West Virginia.
MMWR. Morbidity and mortality weekly report, 35(6), 81-82 (1986-02-14)
M P Oliveira-Neto et al.
Ophthalmology, 107(1), 169-172 (2000-01-27)
To describe American cutaneous leishmaniasis of the eyelids and highlight the main clinical and diagnostic features of lesions, which are rare in this location. Retrospective, noncomparative case series Leishmanin skin test, touch preparations, histopathologic analysis, and culture in appropriate media
Flávio José Dutra de Moura et al.
Toxicology, 243(1-2), 66-74 (2007-11-21)
Tumor necrosis factor-alpha (TNF-alpha) is a mediator of inflammation and has an important role in human and experimental renal diseases. Pentoxifylline (PTX) has been shown to inhibit cytokine synthesis, including TNF-alpha. The aim of the present study was to examine
J Carrió et al.
Experimental parasitology, 95(3), 209-214 (2000-08-31)
The determination of the intrinsic sensitivity of Leishmania strains to pentavalent antimonials in clinical trials, before treatment is begun, is essential in order to avoid failures and to allow alternative drugs to be chosen. A comparative study of SbV activity
Et2SBrSbCl5Br: an effective reagent for direct bromonium-induced polyene cyclizations.
Scott A Snyder et al.
Angewandte Chemie (International ed. in English), 48(42), 7899-7903 (2009-09-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service