SA202
Streptavidin, Peroxidase Conjugated
1 mL Streptavidin, Peroxidase Conjugated for ELISA, Western Blotting & IHC.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
conjugate
peroxidase conjugate
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
shipped in
wet ice
Application
1 mL Streptavidin, Peroxidase Conjugated for ELISA, Western Blotting & IHC.
WORKING DILUTION
The final working dilution is dependent on the specific assay conditions. However, good results can be expected with dilutions in the following ranges:
ELISA: 1/500 - 1/10,000
Immunohistochemistry: 1/40 - 1/1,000
The final working dilution is dependent on the specific assay conditions. However, good results can be expected with dilutions in the following ranges:
ELISA: 1/500 - 1/10,000
Immunohistochemistry: 1/40 - 1/1,000
Physical form
The conjugate is supplied in phosphate buffered saline, pH 7.2, with 0.5% (w/v) bovine serum albumin and 10% (w/w) glycerol as stabilizers, plus added preservative.
Storage and Stability
Store at +2 to +8°C, for up to 6 months.
NOTE: Do not dilute into buffers containing sodium azide, as this inhibits peroxidase.
NOTE: Do not dilute into buffers containing sodium azide, as this inhibits peroxidase.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service