Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PK1109

Sigma-Aldrich

PhosphoDetect Anti-Src Family (pTyr⁴¹⁶) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse, rat, chicken, human, Xenopus

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr416)

Gene Information

human ... SRC(6714)

General description

Protein A and immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~60 kDa Src protein phosphorylated at Tyr416.
Recognizes the ~60 kDa Src protein phosphorylated at Tyr416 in COLO 201 cells and PDGF-treated NIH/3T3 cells. May also recognize other Src family kinases phosphorylated at equivalent sites.
This PhosphoDetect Anti-Src Family (pTyr⁴¹⁶) Rabbit pAb is validated for use in Immunoblotting for the detection of Src Family (pTyr⁴¹⁶).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr⁴¹⁶ phosphorylation site of human Src

Application


Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium HEPES, 100 µg/ml BSA, 50% glycerol, pH 7.5.

Reconstitution

Do not aliquot.

Analysis Note

Positive Control
COLO 201 cells, PDGF-treated NIH/3T3 cells, and breast carcinoma

Other Notes

May also recognize other Src family kinases phosphorylated at equivalent sites. The immunogen sequence is 100% conserved in chicken, rat, and Xenopus, but cross-reactivity has not been tested. For immunoblotting the membrane should be incubated with antibody diluted in 5% BSA, 1X TBS, 0.1% Tween®-20 detergent at 4°C overnight with gentle shaking. Antibody should be titrated for optimal results in individual systems.
Thomas, S.M. and Brugge, J.S. 1997. Annu. Rev. Cell Dev. Biol.13, 513.
Chow, L.M. et al. 1993. Nature365, 156.
Molina, T.J., et al. 1992. Nature357, 161.
Straus, D.B. and Weiss, A. 1992. Cell70, 585.
Yamanashi, Y., et al. 1991. Science251, 192.
Yamanashi, Y., et al. 1989. Proc. Natl. Acad. Sci. USA86, 6538.
Hunter, T. 1987. Cell49, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Barisic et al.
Biochemical pharmacology, 80(4), 439-447 (2010-05-11)
Activation status of Tyr-kinase Src as well as of the transcription factor NFkappaB is a decisive criterion for the onset of cancer and in conveying radio-resistance. While the activation status of Src is Tyr phosphorylation-dependent, NFkappaB activation requires Ser phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service