Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABT187

Sigma-Aldrich

Anti-Kindlin-3 Antibody, clone 181A

clone 181A, from rat

Synonym(s):

Fermitin family homolog 3, Kindlin-3, MIG2-like protein, Unc-112-related protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

181A, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FERMT3(83706)

General description

Kindlin-3 is an intracellular protein encoded by the FERMT3 gene and which binds directly to the β chain cytoplasmic tail of integrin in hematopoietic cells. This protein belongs to a family of adhesion plaque proteins, of which there are three members, which act to activate integrins. Kindlin-3 differs from Kindlins-1 and -2 in that its expression is restricted to hematopoietic systems and is especially high in spleen and megakaryocytes. It is a necessary component for integrin activation and for platelet aggregation. Mutations in FERMT3 and consequently to the encoding of Kindlin-3 protein are causal to LAD-I variant syndrome, an uncommon genetic disorder in which integrin adhesive functions on platelets and leukocytes are disrupted.

Immunogen

His-tagged recombinant protein corresponding to human Kindlin-3.

Application

Anti-Kindlin-3 Antibody, clone 181A is an antibody against Kindlin-3 for use in Western Blotting.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Kindlin-3 in 10 µg of K562 cell lysate.

Target description

~76 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service