Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2246

Sigma-Aldrich

Anti-MerTK Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Mer590, monoclonal

mol wt

calculated mol wt 110.25 kDa

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
activity assay: suitable
flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... MERTK(10461)

Related Categories

General description

Tyrosine-protein kinase Mer (UniProt: Q12866; also known as EC:2.7.10.1, Proto-oncogene c-Mer, Receptor tyrosine kinase MerTK) is encoded by the MERTK (also known as MER) gene (Gene ID: 10461) in human. MerTK is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 21-505), a transmembrane domain (aa 506-526), and a cytoplasmic domain (aa 27-999). Its protein kinase domain is localized to amino acids 587-858. MerTK is highly expressed in testis, ovary, prostate, lung, and kidney tissue. Its expression is not observed in normal B- and T-cells but is expressed in several neoplastic B- and T-cell lines. Overexpression of MerTK has also been reported in non-small cell lung cancer (NSCLC). It is involved in the regulation of cell survival, migration, differentiation, and phagocytosis of apoptotic cells. It transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands, including galectin-3, Tubby protein, Tubby-related protein 1, and GAS6, a member of the Axl subfamily of receptor protein-tyrosine kinases. Ligand binding at the cell surface induces autophosphorylation of MerTK on its intracellular domain that provides docking sites for downstream signaling molecules. Its autophosphorylation on tyrosines 749, 753, and 754 in the activation loop imparts full activity. It is also autophosphorylated on tyrosine 872 that leads to recruitment of downstream partners of the signaling cascade such as phospholipase C 2. Clone Mer590 is shown to rapidly reduce total and surface expression of MerTK and prevents its phosphorylation and downstream signaling. It is also shown to enhance carboplatin induced apoptosis in Colo699 cells and diminish glioblastoma cell migration in vitro. (Ref.: Cummings, CT., et al. (2014). Oncotarget, 5(21); 10434-45; Rogers, AEJ., et al. (2012). Oncogene. 31(38): 4171 4181).

Specificity

Clone Mer590 is a mouse monoclonal antibody that detects human Tyrosine-protein kinase Mer (MerTK). It targets an epitope within the extracellular domain.

Immunogen

Recombinant human MerTK extracellular domain/Fc chimera produced in CHO cells.

Application

Quality Control Testing

Evaluated by Flow Cytometry in A549 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected MerTK in one million A549 cells.

Tested Applications


Western Blotting Analysis: A representative lot detected MerTK in Western Blotting applications (Rogers, A.E.J., et. al. (2012). Oncogene. 31(38):4171-81).

Flow Cytometry Analysis: A representative lot detected MerTK in Flow Cytometry applications. (Cummings, C.T., et. al. (2014). Oncotarget. 5(21):10434-45; Rogers, A.E.J., et. al. (2012). Oncogene. 31(38):4171-81).

Activity Assay: A representative lot of this antibody reduced surface and total MER tyrosine kinase levels in multiple cell lines. (Cummings, C.T., et. al. (2014). Oncotarget. 5(21):10434-45).

ELISA Analysis: A representative lot detected MerTK in ELISA applications (Rogers, A.E.J., et. al. (2012). Oncogene. 31(38):4171-81).

Inhibition Analysis: A representative lot of this antibody inhibited MerTyrosine kinase activity and reduced the migration of Glioblastoma multiforme cells. (Rogers, A.E.J., et. al. (2012). Oncogene. 31(38):4171-81).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MerTK, clone Mer590, Cat. No. MABS2246, is a mouse monoclonal antibody that detects Tyrosine-protein kinase Mer (MerTK) and is tested for use in Activity Assay, ELISA, Flow Cytometry, Inhibition, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service