Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN1594

Sigma-Aldrich

Anti-WWOX Antibody, clone 12G5

clone 12G5, from rat

Synonym(s):

WW domain-containing oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12G5, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WWOX(51741)

General description

WW domain-containing oxidoreductase (WWOX) is also called Fragile site FRA16D oxidoreductase (FOR). WWOX is a putative oxidoreductase that functions as a tumor suppressor and is involved in apoptosis. WWOX is also required for normal bone development, and inhibits Wnt signaling. WWOX is widely expressed, but is most strongly expressed in testis, prostate, and ovary as well as in cancer cell lines. Certain isoforms (Isoform 5 and isoform 6) are thought to only be expressed in tumor cell lines. WWOX is linked to several cancer types like Esophageal cancer (ESCR).

Immunogen

Recombinant protein corresponding to mouse WWOX.

Application

Anti-WWOX Antibody, clone 12G5 | MABN1594 is an antibody against WWOX Antibody for use in Western Blotting, Immunohistochemistry.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected WWOX in 10 µg of mouse 11 day embreyo and mouse E13-14 Brain tissue lysate.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected WWOX in human cerebellum and human cerebral cortex tissue.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected WWOX in mouse cerebellum and mouse hindbrain tissue.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected WWOX in 10 µg of HepG2 cell lysate.

Target description

~46 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service