Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2049

Sigma-Aldrich

Anti-LTBR Antibody, clone 3C8

clone 3C8, from rat

Synonym(s):

Tumor necrosis factor receptor superfamily member 3, Lymphotoxin-beta receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C8, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Ltbr(17000)

General description

Tumor necrosis factor receptor superfamily member 3 (UniProt: P50284; also known as Lymphotoxin-beta receptor, LTBR) is encoded by the Ltbr (also known as Tnfcr, Tnfrsf3) gene (Gene ID: 17000) in murine species. The tumor necrosis factor (TNF) receptor superfamily comprises more than 20 type-I transmembrane proteins. LTBR is a single-pass type I membrane protein that contains 4 TNFR-Cys repeats and serves as a receptor for the heterotrimeric lymphotoxin containing LTA and LTB, and for TNFS14/LIGHT. LTBR plays critical roles in inflammation and lymphoid organogenesis through activation of NF-kB pathway. The binding of heterotrimeric lymphotoxin to LTBR is shown to induce NF-kB activation and cell death in HT29 adenocarcinoma cells. LTBR activation promotes apoptosis via TRAF3 and TRAF5. It is also reported to play a role in the development of lymphoid organs. Mice deficient in the LTBR gene fail to form lymph nodes and lack NK and NK-T cells. In the adults, LTBR signaling is essential for maintaining the splenic architecture, for the integrity of marginal zones, and for compartmentalization of T and B cells.

Specificity

Clone 3C8 detects Tumor necrosis factor receptor superfamily member 3 in NIH3T3 cells.

Immunogen

LT betaR-Fc decoy protein.

Application

Agonist or Inhibitor Analysis: A representative lot detected LTBR in Agonist or Inhibitor applications (Zhu, M., et. al. (2006). J Clin Invest. 116(11):2964-71).

Flow Cytometry Analysis: A representative lot detected LTBR in Flow Cytometry applications (Yi, T., et. al. (2013). Elife. 2:e00757).

Agonist or Inhibitor Analysis: A representative lot detected LTBR in Agonist or Inhibitor applications (Dejardin, E., et. al. (2002). Immunity. 17(4):525-35).

Agonist or Inhibitor Analysis: A representative lot detected LTBR in Agonist or Inhibitor applications (Vondenhoff, M.F., et. al. (2009). J Immunol. 182(9):5439-45).

Agonist or Inhibitor Analysis: A representative lot detected LTBR in Agonist or Inhibitor applications (Yi, T., et. al. (2013). Elife. 2:e00757).

Agonist or Inhibitor Analysis: A representative lot detected LTBR in Agonist or Inhibitor applications (Banks, T.A., et. al. (2005). J Immunol. 174(11):7217-25).
Research Category
Inflammation & Immunology
This rat monoclonal Anti-LTBR, clone 3C8, Cat. No. MABF2049, is tested for use in Flow Cytometry and in functional studies for the detection of Lymphotoxin-beta receptor/LTbeta R.

Quality

Evaluated by Flow Cytometry in NIH/3T3 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected LTBR in one million NIH/3T3 cells.

Target description

44.95 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service