Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE2036

Sigma-Aldrich

Anti-PADI2/PAD2 Antibody, clone DN2

Synonym(s):

PAD2, Peptidylarginine deiminase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DN2, monoclonal

mol wt

observed mol wt ~80 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

rabbit

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
multiplexing: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

Specificity

Clone DN2 is a mouse monoclonal antibody that detects Protein-arginine deiminase 2 (PADI2/PAD2). It targets an epitope within the N-terminal region.

Immunogen

Protein-arginine deiminase (PADI2/PAD2) from rabbit skeletal muscle.

Application

Quality Control Testing

Evaluated by Western Blotting with full-length, recombinant, Human PADI2/PAD2.

Western Blotting Analysis: A 1:500 dilution of this antibody detected full-length, recombinant, Human PADI2/PAD2.

Tested Applications

Immunohistochemistry Applications: A representative lot detected PADI2/PAD2 in Immunohistochemistry applications (Damgaard, D., et al. (2014). J Immunol Methods. 405:15-22).

Western Blotting Analysis: A representative lot detected PADI2/PAD2 in Western Blotting applications (Damgaard, D., et al. (2014). J Immunol Methods. 405:15-22; Bawadekar, M., et al. (2017). J Autoimmun. 80:39-47).

ELISA Analysis: A representative lot detected PADI2/PAD2 in ELISA applications (Damgaard, D., et al. (2014). J Immunol Methods.;405:15-22; Damgaard, D., et al. (2016). Rheumatology (Oxford). 55(5):918-27).

Multiplexing: A representative lot detected PADI2/PAD2 in Multiplexing applications (Vejlstrup, A., et al. (2019). J Inflamm Res. 29:12:137-144).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein-arginine deiminase (UniProt: A0A5F9DB16; also known as PADI2, PAD2) is encoded by the PADI2 gene (Gene ID: 100357368) in rabbit. Protein-arginine deiminases (PAD) catalyze the hydrolysis of peptidyl-arginine to peptidyl-citrulline and alter protein function. Five isoforms of PAD have been described in mammals (PAD1, 2, 3, 4, and 6) that are homologous in their catalytic site but are expressed differently in different cells and tissues and have different physiological roles. PAD2, one of the most abundant PADs has three domains: N-terminal domain (aa 1-114), a central domain (aa 116-274), and a C-terminal domain (aa 285-657). High calcium concentration is reported to be a prerequisite for PAD2 activation. A redox potential maintains its catalytic site in a reduced state. Overexpression of PAD2 has been associated with inflammatory diseases, such as rheumatoid arthritis and neurodegenerative diseases including multiple sclerosis. Higher levels of PAD2 are observed in transformed cell model of breast cancer progression with increasing tumor malignancy and in a murine models of xenograft tumors. Clone DN2 binds to an epitope in the N-terminal region and is ideally suited for quantitative ELISA. (Ref.: Wang, Y., et al. (2021). Int. J. Cancer. 148(2); 267-276; Vejlstrup, A., et al. (2019). J. Inflamm. Res. 12; 137 144; Damgaard, D., et al. (2016). Rheumatology 55(5); 918-927; Damgaard, D., et al. (2014). J. Immunol. Methods. 405; 15-22).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service