Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1074

Sigma-Aldrich

Anti-SMARCA3 (HLTF) Antibody, clone 11-1

clone 11-1, from mouse

Synonym(s):

Helicase-like transcription factor, DNA-binding protein/plasminogen activator inhibitor 1 regulator, HIP116, RING finger protein 80, Sucrose nonfermenting protein 2-like 3, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11-1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HLTF(6596)

General description

Helicase-like transcription factor (UniProt Q14527; also known as DNA-binding protein/plasminogen activator inhibitor 1 regulator, HIP116, RING finger protein 80, Sucrose non-fermenting protein 2-like 3, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3) is encoded by the HLTF (also known as HIP116A, RNF80, SMARCA3, SNF2L3, ZBU1) gene (Gene ID 6596) in human. Helicase-like transcription factor (115 kDa) is a SWI/SNF (mating-type switching/sucrose non-fermenting) protein that uses ATP hydrolysis to remodel chromatin in multiple cellular processes. It is shown to regulate transcription by binding to the SV40 enhancer and the promoters Serpin E1 and HIV-1. It is well expressed in brain, heart, kidney, liver, lung, pancreas, placenta, and skeletal muscle. It was initially characterized for its DNA binding and transcriptional activity, however, later was shown to act as an E3 ubiquitin ligase and was implicated in post-replication DNA repair by polyubiquitination of proliferating cell nuclear antigen (PCNA). The inactivation of HIP116 is shown to induce an elevated level of chromosome breaks and fusions in mouse embryonic fibroblasts after treatment with alkylating agents. HIP116 inactivation in transgenic mice is reported to disrupt cell cycle at the G2/M phase.

Ref.: Arcolia, V et al. (2014). BMC Cancer 14, 492.

Specificity

Clone 11-1 detected predominantly nuclear HLTF immunoreactivity by immunohistochemistry staining. Clone 11-1 epitope lies within the N-terminal region present in human HLTF (SMARCA3) spliced isoform 1, but not in isoform 2 reported by UniProt (Q14527).

Immunogen

GST-tagged recombinant human HLTF (SMARCA3) N-terminal fragment.

Application

Anti-SMARCA3 (HLTF), clone 11-1, Cat. No. MABE1074, is a highly specific mouse monoclonal antibody that targets HTLF and has been tested in Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected predominantly nuclear HLTF (SMARCA3) immunoreactivity in human cervical cancer and cerebellum tissue sections.

Quality

Evaluated by Immunohistochemistry in human breast cancer tissue section.



Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected predominantly nuclear HLTF (SMARCA3) immunoreactivity in human breast cancer tissue section.

Target description

113.9 kDa (human isoform 1) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service