Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABE1012

Sigma-Aldrich

Anti-E2F-7 Antibody, clone 14C3.2

clone 14C3.2, from mouse

Synonym(s):

Transcription factor E2F7, E2F-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
clone:
14C3.2, monoclonal
application:
ICC
WB
species reactivity:
mouse, human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14C3.2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... E2F7(144455)

General description

Transcription factor E2F7 (UniProt Q96AV8; also known as E2F-7) is encoded by the E2F7 gene (Gene ID 144455) in human. The E2F family of transcription factors (E2F) regulate genes involved in cell proliferation, differentiation and apoptosis. E2F1, 2, 3a function as transcription activators, while other family members (E2F3b, 4, 5, 6, 7, 8) are known as transcription repressors. Upregulated E2F7 is found in tamoxifen-resistant breast cancer cells, where it is responsible for transcriptional repression of miR-15a/16. Exogenous expression of miR-15a/16 or silencing E2F7 re-sensitizes breast cancer cells to tamoxifen-induced cell cycle arrest and apoptosis. Upregulated E2F7 expression is also reported in cutaneous squamous cell carcinomas (CSCC) and and head and neck SCC (HNSCC), where E2F7-dependent Sphk1 overexpression in SCC results in an elevated production of the antiapoptotic phospholipid, sphingosine-1-phosphate (S1P), which in turn invokes anthracycline resistance via activation of the PI3K/AKT pathway.

Specificity

Clone 14C3.2 recognizes an internal epitope of human E2F-7 present in all three spliced isoforms reported by UniProt (Q96AV8).

Immunogen

Epitope: Internal (C-terminal half).
GST-tagged recombinant human E2F-7 internal fragment.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected E2F-7 in A431, HeLa, HepG2 and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-E2F-7 Antibody, clone 14C3.2 is validated for use in Western Blotting, Immunocytochemistry for the detection of E2F-7.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected E2F-7 in 10 µg of HEK293 cell lysate.

Target description

~100 kDa observed. 99.89 kDa (isoform 1; E2F7b), 80.87 kDa (isoform 2; E2F7a), 98.35 kDa (isoform 3) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service