Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1166

Sigma-Aldrich

Anti-iASPP Antibody, clone LX128.5

clone LX128.5, from mouse

Synonym(s):

RelA-associated inhibitor, Inhibitor of apoptosis-stimulating protein of p53, Inhibitor of ASPP protein, NF-kB-interacting protein 1, PPP1R13B-like protein, Protein iASPP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LX128.5, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

RelA-associated inhibitor (UniProt Q8WUF5; also known as Inhibitor of apoptosis-stimulating protein of p53, Inhibitor of ASPP protein, NF-kB-interacting protein 1, PPP1R13B-like protein, Protein iASPP) is encoded by the PPP1R13L (also known as IASPP, NKIP1, PPP1R13BL, RAI) gene (Gene ID 10848) in human. Inhibitor of apoptosis-stimulating protein of p53 (iASPP) belongs to the ASPP family of proteins (ASPP1, ASPP2, and iASPP) characterized by their ankyrin repeats (a.a. 659-691 & 692-724 in iASPP), SH3 domain (a.a. 758-820 in iASPP), and proline-rich (a.a. 54-602 in iASPP) sequences. iASPP is expressed at intercalated discs in post-mitotic cardiomyocytes, where it interacts with desmoplakin and desmin in cardiomyocytes to help maintain the integrity of desmosomes and intermediate filament networks. Desmosomes are anchoring junctions found in cells that endure physical stress such as cardiac myocytes or skin keratinocytes. Mutations of genes coding desmosome components, including IASPP/PPP1R13L, are found in human patients suffering from cardiomyopathy and in animal models of cardiomyopathy. iASPP deficiency caused by spontaneous mutation has been reported to be linked to a lethal autosomal recessive cardiomyopathy in Poll Hereford calves and Wa3 mice. iASSP-deficient mice with exon 8 deletion display features of arrhythmogenic right ventricular cardiomyopathy (ARVC) and die of sudden cardiac death.

Specificity

Clone LX128.5 immunostained intercalated discs in myocardial sections from wild-type, but not iASPP-deficient (iASPPΔ8/Δ8) adult mice (Notari, M., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(9):E973-E981).

Immunogen

Synthetic linear peptide corresponding to a sequence from the N-terminal region of human iASPP.

Application

Anti-iASPP Antibody, clone LX128.5 is an antibody against iASPP for use in Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A representative lot immunostained intercalated discs in myocardial sections from adult wild-type, but not iASPP-deficient (iASPPΔ8/Δ8) mice (Notari, M., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(9):E973-E981).
Note: iASPP is expressed at intercalated discs. To prepare heart tissue sections for iASPP staining, it is important to cut the tissues at an angle parallel to the plane of the intercalated discs.

Quality

Evaluated by Immunohistochemistry in mouse heart tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected iASPP in mouse heart tissue.

Target description

89.09 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service