Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAB5434-C

Sigma-Aldrich

Anti-Oct-1 Antibody, clone YL15, Ascites Free

clone YL15, from mouse

Synonym(s):

POU domain, class 2, transcription factor 1, NF-A1, Oct-1, Octamer-binding protein 1, Octamer-binding transcription factor 1, OTF-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

YL15, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU2F1(5451)
mouse ... Pou2F1(18986)

General description

POU domain, class 2, transcription factor 1 (UniProt P14859; also known as NF-A1, Oct-1, Octamer-binding protein 1, Octamer-binding transcription factor 1, OTF-1) is encoded by the POU2F1 (also known as OCT1, OTF1) gene (Gene ID 5451) in human. Oct-1 belongs to the family of octamer transcription factors characterized by their binding to the ATTTGCAT octamer sequence via their POU domain (a.a. 280-354 of human Oct-1). Oct1 is widely expressed in adult tissues, however, Oct-1 knockdown has little impact on cell growth and viability in culture. Oct-1 controls multiple stem cell phenotypes in normal and tumor cells. Oct-1 promotes resistance to genotoxic and oxidative stresses in stem cells. Loss of Oct-1 antagonizes oncogenic transformation in vitro and tumorigenicity in vivo.

Specificity

Clone YL15 targets the POU-domain linker region (Herr, W., and Cleary, M.A. (1995). Genes Dev. 9(14):1679-1693; Lai, J.S., and Herr, W. (1992). Proc. Natl. Acad. Sci. U. S. A. 89(15):6958-6962) present in all six spliced isoforms of human Oct-1 (UniProt P14859). Clone YL15 does not react with Oct-2, Oct-4 or Oct-6 (Maddox, J., et al. (2012). PLoS Genet. 8(11):e1003048).

Immunogen

Epitope: POU-domain linker region.
Recombinant full-length human Oct-1.

Application

Anti-Oct-1 Antibody, clone YL15, Cat. No. MAB5434-C, is a mouse monoclonal antibody that detects octamer-binding protein 1 (Oct-1) and has been tested for use in immunocytochemistry, immunohistchemistry (Parffin), Immunoflouroesencece, Western blotting, and Chromatin Immunorecipitation.
Immunocytochemistry Analysis: 5.0 µg/mL from a representative lot detected Oct-1 cellular localization in human A431 and HeLa cells by fluorescent immunocytochemistry.
Western Blotting Analysis: A representative lot detected Oct-1, but not Oct-2, Oct-4, or Oct-6 in various human cell lysates and nuclear extracts (Maddox, J., et al. (2012). PLoS Genet. 8(11):e1003048).
Western Blotting Analysis: A representative lot detected Oct-1 in Burkitt lymphoma cells (BJAB and Namalwa), as well as a panel of six primary effusion lymphoma (PEL) cells (Di Bartolo, D.L., et al. (2009). J. Virol. 83(9):4308-4315).
Immunofluorescence Analysis: A representative lot detected Oct-1 immunoreactivity in paraformaldehyde-fixed human and mouse frozen colon tissue sections by fluorescent immunohistochemistry (Maddox, J., et al. (2012). PLoS Genet. 8(11):e1003048).
Immunohistochemistry Analysis: A representative lot detected Oct-1 immunoreactivity in formalin-fixed, paraffin-embedded human colon tissue sections (Maddox, J., et al. (2012). PLoS Genet. 8(11):e1003048).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Oct-1 occupancy at the ORF50 promoter, using stable Oct-2-inducible primary effusion lymphoma (PEL) cell line BC1/Tet-on/Oct-2. The association decreased upon induction of Oct-2 expression and enhanced upon ORF50 transfection (Di Bartolo, D.L., et al. (2009). J. Virol. 83(9):4308-4315).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Oct-1 occupancy at the U6 and U1 promoter regions in exponentially dividing HeLa cells (Zhao, X., et al. (2001). Mol. Cell. 7(3):539-549).
Electrophoretic Mobility Shift Assay (EMSA): A representative lot caused a supershift of radiolabeled OCT1 consensus oligonucleotide-bound complex in the nuclear extracts from Burkitt lymphoma cells BJAB and Namalwa (Di Bartolo, D.L., et al. (2009). J. Virol. 83(9):4308-4315).
Immunocytochemistry Analysis: A representative lot detected Oct-1 cellular localization in human diploid fibroblast MRC-5-derived HuS-L12 cells at various population-doubling levels (PDLs), as well as in the corresponding immortalized IML12–4 cells by fluorescent immunocytochemistry (Imai, S., et al. (1997). Mol. Biol. Cell. 8(12):2407-2419).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Oct-1 from HEK293 cell lysates (Lai, J.S., and Herr, W. (1992). Proc. Natl. Acad. Sci. U. S. A. 89(15):6958-6962).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Oct-1 in 10 µg of HeLa cell lysate.

Target description

~96 kDa observed.

Linkage

Replaces: MAB5434

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service