Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB4331

Sigma-Aldrich

Anti-MELK Antibody, clone 6C1.3

clone 6C1.3, Chemicon®, from mouse

Synonym(s):

Maternal Embryonic Leucine Zipper Kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C1.3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

input

sample type neural stem cell(s)

isotype

IgG2κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MELK(9833)

General description

Melk, (Maternal Embryonic Leucine Zipper Kinase) is a serine-threonine kinase of the snfI/AMPK family that is expressed in developing neuroepithelium, self-renewing neural stem cells and granule cell precursors. It is also considered an important brain tumor marker, being highly expressed in gliobastoma, astrocytomas, and medulablastomas.

Immunogen

MELK Recombinant protein.

Application

Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells
This Anti-MELK Antibody, clone 6C1.3 is validated for use in WB for the detection of MELK.
Western blot: Recommended dilution: 1:500 (2 μg/mL)
Expected Molecular Weight: 74.5 kDa
Note: Bands may be present at a higher molecular weight due to glycosolation.

Target description

74.5 kDA; glycosylations may result in higher MW bands in western blot.

Physical form

Format: Purified
Purified immunoglobulin in PBS, pH 7.0. Contains 0.1% sodium azide.

Storage and Stability

1 year at 2-8 °C from date of shipment

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lieve Verlinden et al.
The Journal of biological chemistry, 280(45), 37319-37330 (2005-09-08)
1,25-Dihydroxyvitamin D(3) (1,25(OH)(2)D(3)) has potent antiproliferative effects characterized by a hampered G(1)/S transition. cDNA microarrays were used to monitor expression of 21,492 genes in MC3T3-E1 mouse osteoblasts at 1, 6, 12, 24, and 36 h after treatment with 1,25(OH)(2)D(3). Statistical
Mathew C Easterday et al.
Developmental biology, 264(2), 309-322 (2003-12-04)
The identification of the genes regulating neural progenitor cell (NPC) functions is of great importance to developmental neuroscience and neural repair. Previously, we combined genetic subtraction and microarray analysis to identify genes enriched in neural progenitor cultures. Here, we apply
Ichiro Nakano et al.
The Journal of cell biology, 170(3), 413-427 (2005-08-03)
Maternal embryonic leucine zipper kinase (MELK) was previously identified in a screen for genes enriched in neural progenitors. Here, we demonstrate expression of MELK by progenitors in developing and adult brain and that MELK serves as a marker for self-renewing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service