Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1056

Sigma-Aldrich

Anti-Neutrophil Elastase Antibody, clone AHN-10

ascites fluid, clone AHN-10, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AHN-10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SERP1(27230)

Specificity

Reacts specifically with human neutrophil elastase and not with the 3 homologous neutrophil neutral serine proteases, cathepsin G, esterase N and proteinase 3. Inhibits the elastinolytic activity of elastase. Elastase is felt to play an important role in a number of disease states.

Application

Anti-Neutrophil Elastase Antibody, clone AHN-10 detects level of Neutrophil Elastase & has been published & validated for use in IP, RIA & IC.
Immunoprecipitation

Immunocytochemistry

RIA

Optimal working dilutions must be determined by end user.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Storage and Stability

Maintain in convenient aliquots at -20°C or colder for up to 12 months. Avoid thawing and refreezing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Brittany Johnson et al.
Cellular microbiology, 17(5), 648-665 (2014-10-28)
During gonorrhoeal infection, there is a heterogeneous population of Neisseria gonorrhoeae (Gc) varied in their expression of opacity-associated (Opa) proteins. While Opa proteins are important for bacterial attachment and invasion of epithelial cells, Opa+ Gc has a survival defect after
Julia Mayerle et al.
Gastroenterology, 129(4), 1251-1267 (2005-10-19)
Cadherins play an important role in cell-cell contact formation at adherens junctions. During the course of acute pancreatitis, adherens junctions are known to dissociate-a requirement for the interstitial accumulation of fluid and inflammatory cells-but the underlying mechanism is unknown. Acute
The eicosanoid response to high dose UVR exposure of individuals prone and resistant to sunburn.
Anna Nicolaou,Mojgan Masoodi,Karl Gledhill,Ann Katarina Haylett,Anthony John Thody et al.
Photochemical & Photobiological Sciences : Official Journal of the European Photochemistry Association and the European Society for Photobiology null
Severely photosensitive psoriasis: a phenotypically defined patient subset.
Kirsty J Rutter,Rachel E B Watson,Lindsey F Cotterell,Thomas Brenn et al.
The Journal of Investigative Dermatology null
K M Skubitz et al.
Journal of leukocyte biology, 44(3), 158-165 (1988-09-01)
Neutrophil proteases are believed to play a role in the pathogenesis of a variety of human diseases. While many studies of proteases in models of disease have focused on elastase, neutrophils contain several proteases some of which share a high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service