Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1012-I

Anti-Alkaline Phosphatase Antibody, E. coli

new

Synonym(s):

EC:3.1.3.1, JW0374, alkaline phosphomonoesterase, b0383, glycerophosphatase, phosphomonoesterase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N/A, monoclonal

mol wt

calculated mol wt 49.44 kDa
observed mol wt ~50 kDa

purified by

using protein G

species reactivity

E. coli

packaging

antibody small pack of 100

technique(s)

western blot: suitable

isotype

IgG2bκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Specificity

This mouse recombinant monoclonal antibody detects Alkaline Phosphatase from E. coli.

Immunogen

Purified Alkaline Phosphatase from E. coli.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from E. coli.

Western Blotting Analysis: A 1:1,000 Dilution of this antibody detected Alkaline Phosphatase in E. coli lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected Alkaline Phosphatase in Western Blotting application (Yogev, D., et al. (1991). J Bacteriol. 173(6):2035-44; Taw, M.N., et al. (2022). Sci Rep. 12(1):14862).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Alkaline phosphatase (UniProt: P00634; also known as EC:3.1.3.1, APase) is encoded by the phoA (also known as b0383, JW0374) gene (Gene ID: 945041) in E. coli. Alkaline phosphatase is a dimeric, non-glycosylated metalloenzyme, requiring two zinc ions (Zn²⁺) and one magnesium ion (Mg²⁺) per subunit for its activity and structural stability. The active site of Alkaline phosphatase includes a serine residue that plays a crucial role in its catalytic mechanism. The primary function of E. coli Alkaline phosphatase is to catalyze the hydrolysis of phosphate monoesters, resulting in the release of an inorganic phosphate and an alcohol. The enzyme is involved in various processes, including phosphate metabolism and regulation. Alkaline phosphatase is also known for its ability to transfer phosphate groups to acceptor molecules, which is key in its role in transphosphorylation reactions. Alkaline Phosphatase has a wide range of applications in molecular biology. It is used as a reporter enzyme in various assays, including ELISA and Western blotting. During molecular cloning, it is commonly used to dephosphorylate the 5′ ends of DNA fragments to prevent self-ligation during cloning procedures.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service