Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HG00C144

Millipore

Violet Red Bile MUG Agar

pH 7.4±0.2 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

composition

Agar, 13.0 g/L
Bile Salts No 3, 1.5 g/L
Brain Heart Infusion, 7.0 g/L
Crystal Violet, 0.002 g/L
di-Sodium Hydrogen Phosphate, 1.0 g/L
Lactose, 9.0 g/L
MUG, 0.1 g/L
Neutral Red, 0.03 g/L
Peptone, 4.0 g/L
Sodium chloride, 4.5 g/L

packaging

poly bottle of 12 × 500 g (HG00C144.500)

pH

7.4±0.2 (25 °C)

storage temp.

15-25°C

Application

A selective, fluorogenic medium for the simultaneous detection and enumeration of Escherichia coli and total coliforms in water, milk and food products.

Preparation Note

Suspend 40g in 1 litre demineralised water. Stand for 15 minutes. Boil whilst stirring until completely dissolved. Cool to 45-50°C. Mix well and pour plates. Do not Autoclave

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service