Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB255F

Sigma-Aldrich

Milli-Mark® Anti-Mouse IL-17 FITC Antibody, clone TC11-18H10.1

clone TC11-18H10.1, Milli-Mark®, from rat

Synonym(s):

Cytotoxic T-lymphocyte-associated antigen 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

TC11-18H10.1, monoclonal

species reactivity

mouse

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Il17A(16171)

General description

IL17 is produced primarily by activated CD4+ T cells and induces a variety of target cells to secrete inflammatory cytokines, including IL1beta, IL6, TNFalpha, IFNgamma and granulocyte colony-stimulating factor, as well as synergizing with other inflammatory cytokines and agonists. The role of IL17 in bone metabolism has become a focus area for studies into Rheumatoid Arthritis, in which levels of IL17are significantly increased and may contribute to bone destruction.

Specificity

Antibody recognizes Interleukin-17A.

Immunogen

Recombinant protein corresponding to mouse Il17a

Application

Milli-Mark Anti-Mouse IL-17 FITC Antibody, clone TC11-18H10.1 detects level of Mouse IL-17 & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by flow cytometry using stimulated EL-4 cells

Target description

17 kDa Calculated

Physical form

Antigen Affinity Purified
Purified rat monoclonal IgG1κ conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
PMA and Ionomycin stimulated EL-4 cells.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service