Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DR1122

Sigma-Aldrich

Anti-KLF10 Mouse mAb (4G9)

liquid, clone 4G9, Calbiochem®

Synonym(s):

Anti-Kruppel-like Factor 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G9, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF10(7071)

General description

Purified mouse monoclonal antibody. Recognizes the ~45 kDa KLF10 protein.
Recognizes the ~45 kDa KLF10 protein in K562 and HeLa cells.
This Anti-KLF10 Mouse mAb (4G9) is validated for use in Immunoblotting, Immunocytochemistry for the detection of KLF10.

Immunogen

A recombinant polypeptide corresponding to amino acids of 1-110 human KLF10, expressed as a GST fusion protein

Application



Immunoblotting (1-5 g/ml)Immunocytochemistry (10 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
K562 cells, HeLa cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service