Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABT180

Sigma-Aldrich

Anti-CDCP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

CUB domain-containing protein 1, Membrane glycoprotein gp140, Subtractive immunization M plus HEp3-associated 135 kDa protein, SIMA135, Transmembrane and associated with src kinases, CD318

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDCP1(64866)

General description

CUB domain-containing protein 1 (CDCP1) is also called Membrane glycoprotein gp140, Subtractive immunization M plus HEp3-associated 135 kDa protein (SIMA135), Transmembrane and associated with src kinases and CD_antigen 318. CDCP1 is part of the tetraspanin web involved in tumor progression and metastasis. CDCP1 may also be involved in cell adhesion, cell matrix association and regulation of anchorage versus migration or proliferation versus differentiation via phosphorylation status. CDCP1 is a possible marker for leukemia diagnosis, and for subsets of immature hematopoietic stem cell. CDCP1 is highly expressed in mitotic cells, skeletal muscle, colon, many human tumor cell lines, colorectal cancer, breast carcinoma and lung cancer. CDCP1 may also be found in cells phenotypically similar to mesenchymal stem cells and neural stem cells.

Specificity

Other Homologies: Mouse and Rat (77% sequence homology).
This antibody recognizes the cytoplasmic domain of CDCP1.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human CDCP1.

Application

Anti-CDCP1 Antibody is a highly specific rabbit polyclonal antibody, that targets CUB domain-containing protein 1 & has been tested in western blotting & IHC (Paraffin).
Western Blotting Analysis: 2 µg/mL of this antibody detected CDCP1 in a panel of human tumor tissue lysates.

Immunohistochemistry Analysis: A 1:100 and a 1:500 dilution from a representative lot detected CDCP1 in human breast carcinoma sections.

Quality

Evaluated by Western Blotting in human pancreas tumor tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected CDCP1 in 10 µg of human pancreas tumor tissue lysate.

Target description

~135 kDa and ~70 kDa observed. Uniprot describes 3 isoforms at 92 kDa, 72 kDa, and 38 kDa, due to alternative splicing. The full length and cleaved form of CDCP1 have been observed at ~135 kDa and ~70 kDa (Casar, B., et al. (2012). Oncogene 31(35):3924-3938.). Uncharacterized band(s) may be observed in some cell lysates.

Analysis Note

Control
Human pancreas tumor tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service