Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABS1591

Sigma-Aldrich

Anti-SR-alpha Antibody

serum, from rabbit

Synonym(s):

Signal recognition particle receptor subunit alpha, Docking protein alpha, DP-alpha, SR-alpha, SRalpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IP
WB
species reactivity:
canine, rat, human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

canine, rat, human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SRPR(6734)

General description

Signal recognition particle receptor subunit alpha (UniProt P08240; also known as Docking protein alpha, DP-alpha, SR-alpha, SRalpha) is encoded by the SRPR gene (Gene ID 6734) in human. Secretory proteins typically possess N-terminal hydrophobic signal sequences that direct them to the endoplasmic reticulum (ER) membrane, where they are translocated into the ER lumen for subsequent vesicular transport. Signal recognition particle (SRP) delivers target nascent secretory proteins, while they are still being synthesized, together with the associated ribosome to the ER protein translocon (Sec61 complex) via the interaction with SRP receptor (SR) on the ER membrane. SR is a heterodimer composed of a 70 kDa peripheral membrane protein (SR alpha subunit or SRα) and a 30 kDa singlel-trans membrane protein (SR beta subunit or SRβ). SRα contains an N-terminal SRX domain that mediates its association with SRβ and a C-terminal region which has high homology to the NG domain of SRP54, including a functional GTPase domain. SRβ also possesses a GTPase domain, which interacts with SRα SRX domain only when SRα is in the GTP bound state. SRα forms the major interaction interface with SRP.

Specificity

Expected to react with both spliced isoforms of human SR-alpha reported by UniProt (P08240).

Immunogen

Epitope: Internal (N-terminal half).
KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human SR-alpha.

Application

Immunoprecipitation Analysis: 3 µL from a represenative lot immunoprecipitated SRP receptor alpha subunit (SRα) from canine pancreas tissue rough microsome CHAP extract (Courtesy of Dr. Christopher V. Nicchitta, Duke University Medical Center, Durham, NC).
Western Blotting Analysis: An 1:5,000 dilution of a representative lot detected SR-alpha in 15 µg HeLa cell lysate (Courtesy of Dr. Christopher V. Nicchitta, Duke University Medical Center, Durham, NC).
Western Blotting Analysis: A representative lot detected SRP receptor alpha subunit (SRα) in the Brij35-sensitive (BrS), but not in the Brij35-resistant (BrR) fraction of HeLa ER membrfane proteins (Jagannathan, S., et al. (2014). J. Biol. Chem. 2014, 289(37):25907-25924).
Western Blotting Analysis: A representative lot detected SRP receptor alpha subunit (SRα) in sodium cholate extract (CE) of ribosome-stripped canine pancreas rough microsome (RM) preparation (Migliaccio, G., et al. (1992). J. Cell Biol. 117(1):15-25).
Research Category
Signaling
Research Sub Category
Vesicular Trafficking
This Anti-SR-alpha Antibody is validated for use in Western Blotting, Immunoprecipitation for the detection of SR-alpha.

Quality

Evaluated by Western Blotting in canine pancreas tissue rough microsome lysate.

Western Blotting Analysis: An 1:500 dilution of this antibody detected SR-alpha in canine pancreas tissue rough microsome lysate.

Target description

~75 kDa observed. Target band size appears larger than the calculated molecular weights of 69.81 kDa (isoform 1) and 66.56 kDa (isoform 2).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service