Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABN326

Sigma-Aldrich

Anti-CAPS-2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Calcium-dependent secretion activator 2, CADPS2, CAPS-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human, mouse, rat
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CAPS2(84698)

General description

CAPS-2 and the related protein CAPS-1 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficient neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. Like CAPS-1, numerous isoforms of CAPS-2 are known to exist. These isoforms show distinct expression patterns in the brain. For example, CAPS2b showed high expression in the developing cerebellum. Furthermore, one version of CAPS-2 mRNA that lacks exon 3 is detected in some autistic patients, suggesting that the differential expression pattern of CAPS-2 is involved in neuronal development.

Specificity

This antibody is predicted to be specific to CAPS-2 and not recognize CAPS-1.

Immunogen

KLH-conjugated linear peptide corresponding to human CAPS-2.

Application

Anti-CAPS-2 detects levels of CAPS-2 proteins & has been published & validated for use in WB & IHC.
Western Blot Analysis: 1 µg/mL from a representative lot detected CAPS-2 in 10 µg/mL of human brain tissue lysate.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected CAPS-2 in rat cerebellum tissue.

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected CAPS-2 in 10 µg/mL of mouse brain tissue lysate.

Target description

~160 kDa observed. Uncharacterized bands may appear at ~40 kDa, ~50 kDa, and ~210 kDa in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijun Feng et al.
Nature communications, 8, 14758-14758 (2017-03-21)
Mutations in chromatin modifier genes are frequently associated with neurodevelopmental diseases. We herein demonstrate that the chromodomain helicase DNA-binding protein 7 (Chd7), frequently associated with CHARGE syndrome, is indispensable for normal cerebellar development. Genetic inactivation of Chd7 in cerebellar granule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service