Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE462

Sigma-Aldrich

Anti-phospho ATM/ATR (Thr1989) Antibody

1.0 mg/mL, from rabbit

Synonym(s):

Serine/threonine-protein kinase ATR, Ataxia telangiectasia and Rad3-related protein, FRAP-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

concentration

1.0 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr1989)

Gene Information

human ... ATM(472)

General description

The Ataxia-Telangiectasia and Rad3 related kinase (ATR) is a nuclear serine/threonine kinase that is part of the "first-response" system to DNA damage induced by a wide variety of factors including double-stranded breaks and replication stress. The localization of ATR to sites of DNA damage is aided by the Replication Protein A (RPA). Phospho - ATR (Ser428) targets a number of proteins including BRCA1, WRN, CHEK1, MCM2, and p53/TP53, and coordinates signaling pathways involved in DNA repair and apoptosis. ATR may also be involved in the phosphorylation of the histone H2A.X after replication stress, an early marker of DNA damage. However, ATR also mediates DNA replication and fork stability in normal cell cycles by activation of Chk1. Defective ATR contributes to Seckel syndrome type 1.

Immunogen

Linear peptide corresponding to human ATM/ATR (Thr1989).

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-phospho ATM/ATR (Thr1989) Antibody is validated for use in Western Blotting for the detection of phospho ATM/ATR (Thr1989).
Western Blotting Analysis: A representative lot detected ATM/ATR (Thr1989) in ATR TR cells (Nam, E.A, et al. (2011). JBC. 286(33):28707–28714).

Quality

Evaluated by Western Blotting in 2mM hydroxyurea treated 293T cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected ATM/ATR (Thr1989) in 10 µg of 2mM hydroxyurea treated 293T cell lysate.

Target description

~300 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyoung Kim et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(12), 3097-3108 (2016-12-21)
Purpose: PARP inhibition (PARPi) has modest clinical activity in recurrent BRCA-mutant (BRCAMUT) high-grade serous ovarian cancers (HGSOC). We hypothesized that PARPi increases dependence on ATR/CHK1 such that combination PARPi with ATR/CHK1 blockade results in increased cell death and tumor regression.Experimental
Thr-1989 phosphorylation is a marker of active ataxia telangiectasia-mutated and Rad3-related (ATR) kinase.
Nam, EA; Zhao, R; Glick, GG; Bansbach, CE; Friedman, DB; Cortez, D
The Journal of Biological Chemistry null
Katarzyna B Leszczynska et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 121(2), 232-238 (2016-11-15)
Esophageal cancer has a persistently low 5-year survival rate and has recently been classified as a cancer of unmet need by Cancer Research UK. Consequently, new approaches to therapy are urgently required. Here, we tested the hypothesis that an ATR
Yannick P Kok et al.
Oncogenesis, 9(10), 88-88 (2020-10-09)
Oncogene-induced replication stress, for instance as a result of Cyclin E1 overexpression, causes genomic instability and has been linked to tumorigenesis. To survive high levels of replication stress, tumors depend on pathways to deal with these DNA lesions, which represent
Haineng Xu et al.
Cell reports. Medicine, 2(9), 100394-100394 (2021-10-09)
CCNE1-amplified ovarian cancers (OVCAs) and endometrial cancers (EMCAs) are associated with platinum resistance and poor survival, representing a clinically unmet need. We hypothesized that dysregulated cell-cycle progression promoted by CCNE1 overexpression would lead to increased sensitivity to low-dose WEE1 inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service