Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1057

Sigma-Aldrich

Anti-acetyl Histone H3 Antibody (Lys 64)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys 64)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci. Histone H3 is reported to be acetylated at Lys64 (H3K64ac) by p300 co-activator. In addition, H3K64ac is found enriched at the transcriptional start sites of active genes, consistent with a transcriptional activation modification opposite of its repressive counterpart H3K64me3 (PMID 24668167). The target specificity of ABE1057 is confirmed with peptide-based epitope analysis by dot blot, using Cat. No. 16-667, AbSurance Histone H3 Antibody Specificity Array, and Cat. No. 16-665, AbSurance Histone H2A, H2B, H4 Antibody Specificity Array. Sequence surrounding the target acetylation site is conserved among human Histone H3 subtypes H3.1, H3.2, H3.3, and H3.3C/H3.5 (UniProt P68431, Q71DI3, P84243, and Q6NXT2, respectively).

Immunogen

Epitope: Acetylated Lys 64
KLH-conjugated linear peptide corresponding to human acetyl Histone H3 (Lys 64).

Application

Anti-acetyl Histone H3 Antibody is an antibody against acetyl Histone H3 for use in Western Blotting, Immunocytochemistry, Dot Blot.
Immunocytochemistry Analysis: 2.0 µg/mL from a representative lot detected acetyl Histone H3 (Lys 64) in HeLa, A431, and NIH/3T3 cells.
Dot Blot Specificity Analysis: Histone peptides with various modifications were probed with acetyl Histone H3 (Lys 64) using a 1:250 dilution.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in sodium butyrate treated HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected acetyl Histone H3 (Lys 64) in 10 µg of sodium butyrate treated HeLa cell lysate.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Martire et al.
Nature genetics, 51(6), 941-946 (2019-06-04)
The histone variant H3.3 is enriched at enhancers and active genes, as well as repeat regions such as telomeres and retroelements, in mouse embryonic stem cells (mESCs)1-3. Although recent studies demonstrate a role for H3.3 and its chaperones in establishing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service