Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABD28

Sigma-Aldrich

Anti-Rfx6 Antibody

serum, from rabbit

Synonym(s):

DNA-binding protein RFX6, Regulatory factor X 6, Regulatory factor X domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rfx6(320995)

General description

Rfx6 (regulatory factor X 6) is a winged-helix transcription factor that is essential for development and seems to be located exclusively in the pancreatic islets. It has been associated with endoderm development, specifically, and the differentiation of islet cells. Since insulin production happens in the beta-cells of pancreatic islets, Rfx6 may be used clinically to treat diabetes. Recently, Rfx6 function has been found to change over time and gets restricted to endocrine cells at later stages. Mitchell-Riley syndrome is thought to stem from a Rfx6 mutation.

Immunogen

GST-tagged recombinant protein corresponding to mouse Rfx6.

Application

Anti-Rfx6 Antibody is a Rabbit Polyclonal Antibody for detection of Rfx6 also known as DNA-binding protein RFX6, Regulatory factor X 6 & has been validated in IHC.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Immunohistochemistry in mouse embryo pancreas tissue.

Immunohistochemistry Analysis: 1:400 dilution of this antibody detected Rfx6 in mouse embryo pancreas tissue.

Target description

102 kDa calculated. Additional isoforms at 59 and 73 kDa have also been reported.

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse embryo pancreas tissue

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Lu et al.
Journal of molecular endocrinology, 66(2), 129-140 (2020-12-23)
RFX6 transcription factor is believed to play a central role in directing cell development of insulin-producing pancreatic islet. RFX6 homozygous mutations cause syndromic neonatal diabetes with hypoplastic pancreas. However, RFX6 heterozygous mutations cause maturity-onset diabetes of the young (MODY) with
Transcriptional regulatory factor X6 (Rfx6) increases gastric inhibitory polypeptide (GIP) expression in enteroendocrine K-cells and is involved in GIP hypersecretion in high fat diet-induced obesity.
Suzuki, K; Harada, N; Yamane, S; Nakamura, Y; Sasaki, K; Nasteska, D; Joo, E; Shibue et al.
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service