Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC1434

Sigma-Aldrich

Anti-UVRAG-FS mutant

from rabbit

Synonym(s):

UV radiation resistance-associated gene protein, p63, UVRAG-FS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... UVRAG(7405)

General description

UV radiation resistance-associated gene protein (UniProt: Q9P2Y5; also known as p63) is encoded by UVRAG gene (Gene ID: 7405) in human. UVRAG is a key autophagic tumor suppressor that is shown to be mutated in several common human cancers. UVRAG as a critical regulator of intracellular membrane trafficking, autophagy, and chromosomal stability. It contains four functional domains, a proline-rich domain, a lipid-binding C2 domain, a Beclin1-binding coiled-coil domain (CCD) and a C-terminal domain. UVRAG is truncated by a frameshift (FS) mutation in colorectal cancer cells. UVRAG-FS is shown to abolish the normal functions of UVRAG, including autophagy, in a dominant-negative manner. Ectopic expression of UVRAG-FS in both embryonic stem cells and cancer cells results in extensive centrosome amplification and concomitant aneuploidy. UVRAG-FS can also enhance the sensitivity to anticancer agents, such as 5-fluorouracil (5-FU), oxaliplatin, and irinotecan. Expression of UVRAG-FS is reported to trigger colorectal cancer metastatic spread through Rac1 activation and epithelial-to-mesenchymal transition, independently of autophagy. UVRAG-FS metastatic tumors display higher levels of p62 than primary tumors, which indicates suppressed autophagy. (Ref.: He, S., et al. (2015). Nat Commun. 6: 7839).

Specificity

This polyclonal antibody targets an internal sequence in the N-terminal half and detects UVRAG-FS by Western blotting.

Immunogen

A frameshift (FS)-UVRAG derived linear peptide.

Application

Anti-UVRAG-FS mutant, Cat. No. ABC1434, detects FS mutant UV radiation resistance-associated gene protein. Validated for use in Immunoprecipitation and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected UV radiation resistance-associated gene protein in Wild-type (WT) and FS mutant UVRAG protein expression in MSS and MSI CRC cell lines (Data courtesy of Dr Liang Laboratory).

Western Blotting Analysis: A representative lot detected Wild-type (WT) and FS mutant UVRAG protein expression in MSS and MSI CRC cell lines. (He, S., et. al. (2015). Nat Commun. 6:7839).

Immunoprecipitation Analysis: A representative lot detected Wild-type (WT) and FS mutant UVRAG protein expression in MSS and MSI CRC cell lines. (He, S., et. al. (2015). Nat Commun. 6:7839).

Quality

Evaluated by Western Blotting in WT/ Mutant UVRAG-FS lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected UV radiation resistance-associated gene protein in Wild-type (WT) and FS mutant UVRAG protein expression in microsatellite stable (MSS) and microsatellite instable (MSI) colorectal cancer cell lines.

Target description

~32 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Ammonium Sulfate
Format: Semi-Purified
Rabbit polyclonal antibody 1XPBS with 0.05% azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service