Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5848

Sigma-Aldrich

Anti-GABA B Receptor R2 Antibody, a.a. 42-54 rat

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, rat
technique(s):
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABBR2(9568)

General description

The inhibitory neurotransmitter, GABA, acts through two ligand-gated channels, GABA(A) and GABA© receptors and GABA(B), the latter of which acts through a G protein mechanism. GABA(B)R2 is one of the two G-protein coupled receptors for GABA(B) that acts as a heteromeric assembly with GABA(B)R1.

Specificity

GABAB Receptor R2 subunit, N-terminus. No reactivity with GABABR1 has been detected.

Immunogen

Epitope: a.a. 42-54 rat
peptide residues 42-54 of the rat 105 kDa GABA(b)R2 protein

Application

Immunoblotting: 1:500-1:1,000. Membrane preparations with proteinase inhibitors are recommended. Whole rat brain membrane preparations. The antibody detects a ~105kDa band under standard western conditions. The immunogen used is 100% conserved in human by sequence analysis; other species have not been examined for reactivity or homology.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-GABA B Receptor R2 Antibody, a.a. 42-54 rat is validated for use in WB for the detection of GABA B Receptor R2.

Physical form

Format: Purified
Purified immunoglobulin. Liquid. Buffer = PBS containing 0.02% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential trafficking of AMPA and NMDA receptors during long-term potentiation in awake adult animals.
Williams, JM; Guevremont, D; Mason-Parker, SE; Luxmanan, C; Tate, WP; Abraham, WC
The Journal of Neuroscience null
Distribution and function of GABAB receptors in spider peripheral mechanosensilla.
Panek, I; Meisner, S; Torkkeli, PH
Journal of Neurophysiology null
Immunohistochemical and immunoblot analysis of gamma-aminobutyric acid B receptor in the prefrontal cortex of subjects with schizophrenia and bipolar disorder.
Masanori Ishikawa, Katsuyoshi Mizukami, Masahiko Iwakiri, Takashi Asada
Neuroscience Letters null
Functional expression of the GABAB receptor in human airway smooth muscle.
Osawa, Y; Xu, D; Sternberg, D; Sonett, JR; D'Armiento, J; Panettieri, RA; Emala, CW
American Journal of Physiology. Lung Cellular and Molecular Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service