Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3500P

Sigma-Aldrich

Anti-Anion Exchanger 1 Antibody

Chemicon®, from rabbit

Synonym(s):

AE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes rat Anion Exchanger 1 (AE1). The immunogen shows no significant sequence homology with other AE or other proteins.

SPECIES REACTIVITIES: The immunogen sequence is 90% conserved in mouse, 75% in human and 70% in bovine. Reactivity with other species has not been confirmed.

Immunogen

A 20 amino acid peptide sequence near the N-terminus of rat AE1 (Kudrycki et al. 1989; Kopito et al. 1985; Raida et al. 1989; Lux et al. 1989; Tanner et al. 1988).

Application

Anti-Anion Exchanger 1 Antibody detects level of Anion Exchanger 1 & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 μg/mL using ECL. AE1 has a molecular weight of approximately 90-100 kDa. It aggregates easily forming dimers and multimers. It may also be proteolytically processed to smaller fragments of 60, 40 and 20 kDa.



ELISA: 1-10 μg/mL using 1 μg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Gröger et al.
The Journal of biological chemistry, 285(19), 14467-14474 (2010-02-27)
Maintenance of ion concentration gradients is essential for the function of many organs, including the kidney, the cornea, and the inner ear. Ion concentrations and fluid content in the cornea are regulated by endothelial cells that separate the collagenous avascular
Nicole Gröger et al.
Human molecular genetics, 21(5), 1025-1036 (2011-11-16)
The human SLC4A5 gene has been identified as a hypertension susceptibility gene based on the association of single nucleotide polymorphisms with blood pressure (BP) levels and hypertension status. The biochemical basis of this association is unknown particularly since no single
Sekiya Shibazaki et al.
Human molecular genetics, 17(11), 1505-1516 (2008-02-12)
Polycystic kidney disease (ADPKD) results from failure of the kidney to properly maintain three-dimensional structure after loss of either polycystin-1 or -2. Mice with kidney selective inactivation of Pkd1 during embryogenesis develop profound renal cystic disease and die from renal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service