Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AB3383-I

Sigma-Aldrich

Anti-UTF1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

UTF1, Undifferentiated embryonic cell transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UTF1(8433)

General description

Undifferentiated embryonic cell transcription factor 1 is a nucleus protein encoded by the UTF1 gene in human and functions as a transcriptional coactivator of ATF2. UTF1 is reported to interact with the ATF-2 metal binding motif via the leucine zipper motif within its conserved domain 2 (CD2) and both CD1 & CD2 of UTF1 and also independently mediates association with TATA-binding protein-containing complex. (PMID 9749258)

Specificity

Other homologies: mouse, rat 95%.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human UTF1 near N-terminus.

Application

Anti-UTF1 Antibody is an antibody against UTF1 for use in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected UTF1 in mouse kidney and rat liver tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in mouse embryonic stem cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected UTF1 in 10 µg of mouse embryonic stem cell lysate.

Target description

~48 kDa observed. UTF1, a novel transcriptional coactivator expressed in pluripotent embryonic stem cells and extra-embryonic cells (Okuda, A., et al. (1998). EMBO. 17(7):2019–2032).

Linkage

Replaces: AB3383

Physical form

Affinity
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UTF1, a novel transcriptional coactivator expressed in pluripotent embryonic stem cells and extra-embryonic cells.
Okuda, A; Fukushima, A; Nishimoto, M; Orimo, A; Yamagishi, T; Nabeshima, Y; Kuro-o et al.
The Embo Journal null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service