Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AB2989

Sigma-Aldrich

Anti-Metadherin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Astrocyte elevated gene-1 protein, Lysine-rich CEACAM1 co-isolated protein, Metastasis adhesion protein, astrocyte elevated gene-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
human, mouse
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
1

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTDH(92140)

General description

Metadherin is a type 2 transmembrane protein who′s overexpression was first described in breast cancers. It plays an important role in the metastasis of breast cancers into the lungs as a secondary site of development. Antibodies reactive to the lung-homing domain of metadherin and siRNA-mediated knockdown of metadherin expression in breast cancer cells inhibited experimental lung metastasis, indicating that tumor cell metadherin mediates localization at the metastatic site (Brown, 2009).

Specificity

The antibody recognizes Metadherin at and around the homing domain.

Immunogen

Epitope: Homing domain
Recombinant GST-tagged proteins corresponding at or around the lung homing domain of mouse Metadherin.

Application

Anti-Metadherin Antibody is an antibody against Metadherin for use in WB & IC.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western blot Analysis: 0.25 µg/ml of this antibody detected Metadherin in 10 µg of HEK293, HepG2, and Jurkat cell lysates.

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Metadherin in NIH/3T3 and HeLa cells.

Quality

Evaluated by Western blot in HeLa cell lysate.

Western blot Analysis: 0.25 µg/ml of this antibody detected Metadherin in 10 µg of HeLa cell lysate.

Target description

~ 75 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jodi E Eipper-Mains et al.
RNA (New York, N.Y.), 17(8), 1529-1543 (2011-06-29)
MicroRNAs (miRNAs) are small RNAs that modulate gene expression by binding target mRNAs. The hundreds of miRNAs expressed in the brain are critical for synaptic development and plasticity. Drugs of abuse cause lasting changes in the limbic regions of the
Sung-Suk Suh et al.
PLoS genetics, 10(10), e1004652-e1004652 (2014-10-24)
Metastasis is the principal cause of cancer death and occurs through multiple, complex processes that involve the concerted action of many genes. A number of studies have indicated that the Fragile Histidine Triad (FHIT) gene product, FHIT, functions as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service