Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AB2266

Sigma-Aldrich

Anti-VPAC2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

vasoactive intestinal peptide receptor 2, Helodermin-preferring VIP receptor, Pituitary adenylate cyclase-activating polypeptide type III receptor, vasoactive intestinal polypeptide receptor 2, PACAP type III receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

pig (based on 100% sequence homology), bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VIPR2(7434)

General description

Vasoactive intestinal polypeptide receptor 2 (VPAC2), also known as PACAP type III receptor or PACAP-R3 for short, is a class B G-protein-coupled receptor. Three distinct PACAP receptors have been identified. Of the three, only PACAP-3 has been found expressed in Sartoli cells of the testis and is coupled to G and adenylyl cyclase, thus having a potential influence on the production of cAMP. In the immune system, PACAP-R3 is expressed on stimulated CD4 T cells, and binding of T cell-derived VIP to-PACAP-R3 induces a shift toward the Th2 pathway. In addition, PACAP-R3 is an essential regulator of the circadian pacemaker of the hypothalamic suprachiasmatic nuclei (SCN).

Specificity

Other homologies: mouse (84% sequence homology).
This antibody recognizes VCAP2 (PACAP-R3) at the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to the extracellular domain of human VPAC2 (PACAP-R3).

Application

Anti-VPAC2 Antibody is an antibody against VPAC2 for use in WB, IH(P).
Immunohistochemistry Analysis: 1:400 dilution from a previous lot detected VPAC2 in normal human cerebral cortex and T cells inside the mantle zone of normal human tonsil tissues.

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected VPAC2 on 10 µg of Jurkat cell lysate.

Target description

~52 kDa observed. An uncharacterized band appears at ~39 kDa in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liwen Chen et al.
Investigative ophthalmology & visual science, 59(7), 2848-2860 (2018-07-20)
To investigate the roles of vasoactive intestinal peptides (VIPs) in regulating the morphology and F-actin distribution of Schlemm's canal (SC) of rat eyes. Chronic intraocular pressure (IOP) hypertension models with episcleral venous cauterization (EVC) were treated with topical VIP or
Xiaoqin Yan et al.
Investigative ophthalmology & visual science, 61(6), 45-45 (2020-06-24)
A previous study reported that vasoactive intestinal peptide (VIP) can regulate the cytoskeleton of Schlemm's canal (SC) endothelium and expand the SC lumen in a rat glaucoma model. In this study, we aimed to investigate the molecular mechanism of VIP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service