Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15640

Sigma-Aldrich

Anti-LMO4 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LMO4(8543)

Specificity

LMO4. By Western blot the antibody recognizes a band at ~18 kDa on MCF-7, human breast carcinoma cell line lysate.

Immunogen

Synthetic peptide.

Application

Detect LMO4 using this Anti-LMO4 Antibody validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuroregenerative Medicine
Western blot: 1-5 μg/mL on MCF-7, human breast carcinoma cell line lysate.

Optimal working dilutions must be determined by end user.

Target description

18 kDa

Physical form

ImmunoAffinity Purified
Liquid in PBS with 0.1% sodium azide.

Storage and Stability

6 months at 4°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service