Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15300

Sigma-Aldrich

Anti-NECL-1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CADM3(57863)

General description

Nectins are immunoglobulin superfamily adhesion molecules that participate in the organization of epithelial and endothelial junctions. Human and mouse NECL1 share 87.3% identity at the amino acid level (1). NECL1 contains an ectodomain made of three immunoglobulin-like domains, and a cytoplasmic region homologous to those of glycophorin C and contactin-associated protein. NECL1 predominantly expressed in the central nervous system, mainly in neuronal cell bodies in a variety of brain regions including the cerebellum, cerebral cortex and hippocampus. NECL1 localizes to the cell-cell junctions and recruits protein 4.1N to the plasma membranes through its C-terminus, thus may regulate the function of the cell-cell junction.

Specificity

NECL-1

Immunogen

Synthetic Peptide

Application

Detect NECL-1 using this Anti-NECL-1 Antibody validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism
Western Blot Analysis: NECL-1 can be detected at 1/500 dilution. Testing at further dilution was not conducted.

Immunohistochemistry: 1:50 to 1:1000 straight dilution of Cat. # AB15301 was tested on human medulla oblongata tissue.

Optimal working dilutions must be determined by the end user.

Target description

43 kDa

Physical form

ImmunoAffinity Purified
Purified Antibody in PBS with 0.1% NaN3

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
Tested on Adult mouse brain lysate

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service