Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AB10339

Sigma-Aldrich

Anti-UGT1a1 Antibody

serum, Chemicon®

Synonym(s):

UDP glucuronosyltransferase 1 family, polypeptide A1.

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

General description

The 9 UDP-glucuronosyltranferases (UGTs) encoded by the UGT1 locus in humans are key enzymes in the metabolism of most drugs as well as endogenous substances such as bile acids, fatty acids, steroids, hormones, neurotransmitters, and bilirubin.

Immunogen

Epitope: AA W42 – K80
Recombinant UGT1a1

Application

Detect UGT1a1 using this Anti-UGT1a1 Antibody validated for use in WB.

Quality

Routinely evaluated by immunoblot.

Target description

60 kDa

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Roos et al.
Toxicology, 284(1-3), 42-53 (2011-04-05)
PCB 180 (2,2',3,4,4',5,5'-heptachlorobiphenyl) is a persistent and accumulating polychlorinated biphenyl abundantly present in food and the environment. In this study, we used highly purified PCB 180 (dioxinlike impurities: 2.7 ng TEQ(WHO)/g PCB 180) in a 28-day toxicity study in young

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service