Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41822

Sigma-Aldrich

4-Chloro-α-methylstyrene

for synthesis

Synonym(s):

4-Chloro-α-methylstyrene, 4-Isopropenylchlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Cl
CAS Number:
Molecular Weight:
152.62
MDL number:
UNSPSC Code:
12352101
EC Index Number:
216-984-1

Assay

≥96.0% (GC)

form

liquid

transition temp

flash point 73 °C

density

1.072 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H9Cl/c1-7(2)8-3-5-9(10)6-4-8/h3-6H,1H2,2H3

InChI key

WQDGTJOEMPEHHL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.070 - 1.074
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

73 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service