Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41378

Sigma-Aldrich

α-Benzoin oxime

for synthesis

Synonym(s):

α-Benzoin oxime, Cuprone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2
CAS Number:
Molecular Weight:
227.26
MDL number:
UNSPSC Code:
12352126
EC Index Number:
207-127-2

form

powder

mp

153-154 °C

storage temp.

2-30°C

InChI

1S/C14H13NO2/c16-14(12-9-5-2-6-10-12)13(15-17)11-7-3-1-4-8-11/h1-10,14,16-17H/b15-13+

InChI key

WAKHLWOJMHVUJC-FYWRMAATSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service