Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20630

Sigma-Aldrich

Hexachloroacetone

for synthesis

Synonym(s):

Hexachloroacetone, Hexachloro-2-propanone, Perchloroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl6O
CAS Number:
Molecular Weight:
264.75
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-129-5

Assay

≥98.0% (GC)

form

liquid

potency

2980 mg/kg LD50, skin (Rabbit)

mp

-3 °C

density

1.74 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3Cl6O/c4-2(5,6)1(10)3(7,8)9

InChI key

DOJXGHGHTWFZHK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.740 - 1.742
Identity (IR): passes test
Color (visual): colorless to yellowish to pink

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service