Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18414

Sigma-Aldrich

1,1,3,3-Tetramethyldisiloxane

for synthesis

Synonym(s):

1,1,3,3-Tetramethyldisiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)2SiH)2O
CAS Number:
Molecular Weight:
134.32
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-906-4

vapor pressure

250 hPa ( 20 °C)

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

240 °C

expl. lim.

0.8-62.9 % (v/v)

reaction suitability

reagent type: reductant

bp

70-71 °C/1013 hPa

mp

-78 °C

transition temp

flash point -26 °C

density

0.76 g/cm3 at 20 °C

InChI

1S/C4H12OSi2/c1-6(2)5-7(3)4/h1-4H3

InChI key

KWEKXPWNFQBJAY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.757 - 0.759
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(C)

-9.99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service